Learn More
Thermo Scientific™ MspI (HpaII) (10 U/μL)
Shop All Thermo Scientific ProductsBeskrivelse
Lambda DNA digested with MspI (HpaII), 1.4% agarose, 328 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...C▵C G G...3'
3'...G G C▵C...5'
Conditions for 100% Activity:
- 1X Buffer Tango™: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- MspI is supplied in: 10mM potassium phosphate (pH 7.5 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: completely overlaps —no effect
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Compatible Ends:
- Bsp119I, Bsu15I, Hin1I, Hin6I, MaeII, NarI, Psp1406I, SsiI, TaqI, XmiI
Note:
MspI is an isoschizomer of HpaII. When the external C in the sequence CCGG is methylated, MspI and HpaII cannot cleave. However, unlike HpaII, MspI can cleave the sequence when the internal C residue is methylated.
Tekniske data
Tekniske data
| Concentration | 10 U/μL |
| Methylation Sensitivity | Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive |
| Enzyme | MspI (HpaII) |
| Compatible Buffer | 10X Buffer Tango |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 3,000 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.