Learn More
Thermo Scientific™ O'GeneRuler™ Ready-to-use 100bp Plus DNA Ladder
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
81.50 € - 90.30 €
Specifications
Color | Orange |
---|---|
Concentration | 6X Loading Dye |
Product Type | DNA Molecular Weight Marker/Ladder |
Form | Liquid |
Kit Contents | 6X Orange DNA Loading Dye |
Description
Thermo Scientific™ O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments. O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose gel orange G dye migrates at 50 bp. The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Can be directly loaded and are stable at room temperature for 6 months
- Supplied with loading dye for sample DNA
Specifications
Orange | |
DNA Molecular Weight Marker/Ladder | |
6X Orange DNA Loading Dye |
6X Loading Dye | |
Liquid | |
100 Applications |
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.