Learn More
Thermo Scientific™ O'GeneRuler 1 kb Plus DNA Ladder, Ready-to-Use - 75-20,000 bp
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this 1 kb DNA ladder, premixed with blue and yellow tracking dyes.
Brand:  Thermo Scientific™ SM1343
Product Code. 11511635
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
O'GeneRuler 1 kb Plus DNA Ladder - 75-20,000 bp, ready-to-use is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
O'GeneRuler 1 kb Plus DNA Ladder, ready-to-use is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose, gel orange G dye migrates at 50 bp.
The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
Highlights
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X Loading Dye
Liquid
500 Applications
DNA Molecular Weight Marker/Ladder
6X Orange DNA Loading Dye, 2 x 1.00mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.