missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Random Hexamer Primer

Thermo Scientific Random Hexamer Primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends.

Brand:  Thermo Scientific™ SO142

44.44 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10580091

  • 59.25 € / 120µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Random Hexamer Primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution (24 μg at 100 μM).

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Oligo(dT)18 Primer (Cat. No. SO131)
Anchored Oligo(dT)20 (Cat. No. 12577011)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Primer
Dry Ice
Random Hexamer
cDNA Synthesis
• Random Hexamer Primer (120 μL at 0.2 μg/μL)

Store at –20°C.
0.2 μg/μL
120 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.