Learn More
Thermo Scientific™ Rapid DNA Ligation Kit
Shop All Thermo Scientific ProductsBeskrivelse
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
- Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes
- Convenient—reaction mixture can be used directly for bacterial transformation
Recommended for:
Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning.
Note:
Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Prior to electroporation it is necessary to inactivate T4 DNA ligase (1) by chloroform extraction.
Tekniske data
Tekniske data
| Buffer | 5X Rapid Ligation Buffer |
| Enzyme | DNA Ligase |
| Product Type | Rapid DNA Ligation Kit |
| Quantity | 50 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.