missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RevertAid H Minus Reverse Transcriptase (200 U/μL)

Thermo Scientific RevertAid H Minus Reverse Transcriptase is a recombinant M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to a point mutation in the RNase H domain.

Brand:  Thermo Scientific™ EP0452

518.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11541515

  • 740.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific RevertAid H Minus Reverse Transcriptase is a recombinant M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to a point mutation in the RNase H domain. RevertAid H Minus Reverse Transcriptase does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA, and therefore high yields of full-length cDNA from long templates are obtained. RevertAid H Minus Reverse Transcriptase maintains activity over a wide temperature range (42–50°C).

Features of RevertAid H Minus Reverse Transcriptase include:
• High yields of full-length first strand cDNA up to 13 kb
• Optimum activity at 42–45°C
• Active up to 55°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)

Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Reverse transcription at elevated temperatures to reduce effects of secondary structure
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• RevertAid H Minus Reverse Transcriptase (200 U/μL)
• 5X Reaction Buffer

Store at –20°C.

60 min.
42°C to 45°C
RevertAid H Minus
Dry Ice
First-Strand cDNA
Reverse Transcription
RNA
Stand-alone Enzyme
Reverse Transcription
5 x 10,000 Units
Reduced
200 U/μL
Separate components
Up to 13 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.