missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T3 promoter Sequencing Primer, 17-mer

Product Code. 10420290 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 μM, 60 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10420290

Brand: Thermo Scientific™ SO119

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends.

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. The primers are complementary to the T3 RNA Polymearse promoter region and are supplied as 10 µM aqueous solutions.

Applications
• Sequencing of DNA fragments located downstream from the T3 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R (Cat. No. SD0141), pTZ57R, and pBluescript II

Promoter sequence 5'-d (ATTAACCCTCACTAAAG)-3'

Related products
T3 promoter Sequencing Primer, 24-mer (SO120)
T7 promoter Sequencing Primer, 20-mer (SO118)
SP6 promoter Sequencing Primer, 18-mer (SO116)
SP6 promoter Sequencing Primer, 24-mer (SO117)

Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated

TRUSTED_SUSTAINABILITY

Specifications

Promoter SP6, T3, T7
Product Type Sequencing Primer
Content And Storage T3 promoter Sequencing Primer, 17-mer, 10 μM (60 μL)

Store at –20°C.
Shipping Condition Dry Ice
Concentration 10 μM
Primer T3
Quantity 10 μM, 60 μL
Vector pTZ19R, pTZ57R, pBluescript II
For Use With (Application) Sequencing
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.