Learn More
Thermo Scientific™ T4 DNA Polymerase (5 U/μL)
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Brand:  Thermo Scientific™ EP0062
 View more versions of this product
Product Code. 10531051
  • 225.00 € / 500 units
Explore more special offers
Description
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
  • Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
  • Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
T4 DNA Polymerase
500 units
2 x 1mL of 5X Reaction Buffer