missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T7 DNA Polymerase (10 U/μL)

Catalyze DNA synthesis in the 5'≥3' direction with T7 DNA Polymerase, a template dependent DNA polymerase.

Brand:  Thermo Scientific™ EP0081

67.45 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10529710

  • 96.50 € / 300 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

T7 DNA Polymerase, a template dependent DNA polymerase, catalyzes DNA synthesis in the 5'≥3' direction. It is a highly processive DNA polymerase allowing continuous synthesis of long stretches of DNA. The enzyme also exhibits a high 3'≥5' exonuclease activity towards single- and double-stranded DNA.

Highlights

  • Strong 3'=>5' exonuclease activity, approximately 1000-fold greater than Klenow Fragment (see Reference 1)
  • Active in restriction enzyme buffers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

T7 DNA Polymerase
300 units
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ T7 DNA Polymerase (10 U/μL) > 10μ/μL, 300U

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.