missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T7 promoter Sequencing Primer, 20-mer

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends.

Brand:  Thermo Scientific™ SO118

37.33 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 10598360

  • 56.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. The primers are complementary to the T7 RNA Polymarese promoter region. Primers are supplied as 10 µM aqueous solutions.

Applications
• Sequencing of DNA fragments located downstream from the T7 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R (Cat. No. SD0141), pTZ57R, and pBluescript II

Promoter sequences 5'-d (TAATACGACTCACTATAGGG)-3'

Related products
T3 promoter Sequencing Primer, 17-mer (SO119)
SP6 promoter Sequencing Primer, 24-mer (SO117)
T3 promoter Sequencing Primer, 24-mer (SO120)
SP6 promoter Sequencing Primer, 18-mer (SO116)

Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SP6, T3, T7
T7 promoter Sequencing Primer, 20-mer, 10 μM

Store at –20°C.
10 μM
10 μM
Sequencing
Sequencing Primer
Dry Ice
T7
pTZ19R, pTZ57R, pBluescript II
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.