missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TaaI (HpyCH4III) (10 U/μL)

The TaaI (HpyCH4III) restriction enzyme recognizes ACN^GT sites and cuts best at 65°C in Tango buffer (Isoschizomers: Bst4CI, HpyCH4III).

Brand:  Thermo Scientific™ ER1361

81.15 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10548170

  • 87.50 € / 200 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lambda DNA digested with TaaI (HpyCH4III), 1.4% agarose, 187 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A C N▵G T...3'

3'...T G▵N C A...5'

Conditions for 100% Activity

  • 1X Buffer Tango: 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 65°C
  • To ensure higher efficiency of digestion, perform the cleavage reaction under paraffin oil

Storage Buffer

  • TaaI is supplied in:10mM Tris-HCl (pH7.5 at 25°C), 100mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with TaaI, approximately 90% of the DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — cleavage impaired
  • EcoKI: may overlap — effect not determined
  • EcoBI: may overlap — effect not determined

Compatible Ends

  • Hpy188I

Note:

Incubation at 37°C results in 10% activity.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
TaaI (HpyCH4III)
No
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer Tango
65°C
200 U
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.