missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, recombinant (5 U/μL)

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus.

Brand:  Thermo Scientific™ EP0401

 View more versions of this product

Product Code. 10374980

  • 57.75 € / 100 units

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Features

  • Thermostable—half life is more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • 10X Taq Buffer without Detergent is recommended for microarray experiments.
  • 10X Taq Buffer with (NH4)2SO4 allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Taq DNA Polymerase (5 U/μL), 20 μL
• 10X Taq Buffer with KCl, 0.6 mL
• 10X Taq Buffer with (NH4)2SO4, 0.6 mL
• 25 mM MgCl2, 0.6 mL

Store at -20°C.
Taq DNA Polymerase
Taq DNA Polymerase (Recombinant)
Dry Ice
5 U/μL
No
Separate Components
Low
Standard
100 Units
Standard PCR
1X
3'-A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2798032 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberEP0401
Lot Number2820416 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0401
Lot Number2803178 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0401
Lot Number2820456 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0401
Lot Number2806418 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0401
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Taq DNA Polymerase, recombinant (5 U/μL) > 100 Units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.