Learn More
Thermo Scientific™ TopVision Agarose
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
149.00 € - 576.00 €
Specifications
Product Type | Agarose Gel |
---|---|
Separation Range | 100 to >30000 bp |
Melting Point | 88 °C |
Product Line | TopVision |
Validated Application | Gel Electrophoresis |
Description
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
Specifications
Agarose Gel | |
88 °C | |
Gel Electrophoresis |
100 to >30000 bp | |
TopVision |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.