Learn More
Thermo Scientific™ UTP Solution (100 mM)
Thermo Scientific UTP (uridine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.
Brand: Thermo Scientific™ R0471
25.78 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific UTP (uridine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This highly stable nucleotide is more than 99% pure. UTP can be used in many different molecular biology applications.
Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles
Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• in vitro transcription
Specifications
100 mM | |
UTP Solution, 100 mM (0.25 mL) Store at –20°C. |
|
Unconjugated | |
UTP Solution |
in vitro Transcription, RNA Amplification | |
Dry Ice | |
0.25 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.