Learn More
Thermo Scientific™ Van91I (PflMI) (10 U/μL)
Cut at CCANNNN^NTGG sites with Van91I (PflMI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: AccB7I, BasI, PflMI).
Brand: Thermo Scientific™ ER0712
159.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Lambda DNA digested with Van91I (PflMI), 0.7% agarose, 14 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'..C C A NN N N^N T G G...3'
3'..G G T N^N N NN A C C...5'
Conditions for 100% Activity
- 1X Buffer R: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/ml BSA
- Incubate at 37°C
Storage Buffer
- Van91I is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with Van91I, more than 90% of DNA fragments can be ligated and recut
Methylation Effects
- Dam: never overlaps – no effect
- Dcm: may overlap – blocked
- CpG: never overlaps – no effect
- EcoKI: never overlaps – no effect
- EcoBI: never overlaps – no effect&
Digestion of Agarose-embedded DNA
- Minimum 5 units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Note:
Van91I is blocked by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099).

Specifications
10 U/μL | |
Van91I (PflMI) | |
Yes | |
No | |
Restriction Enzyme |
Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer R | |
37°C | |
2,000 units | |
Traditional Cloning |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.