missing translation for 'onlineSavingsMsg'
Learn More

Ferrocene, 99%, Thermo Scientific Chemicals

Código de producto. 11344949
Click to view available options
Quantity:
50 g
250 g
1 kg

Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name cyclopenta-1,3-diene;iron
SMILES [Fe].c1cccc1.c1cccc1

Especificaciones

Melting Point 173°C to 174°C
Color Yellow to Orange
Boiling Point 249°C
Odor Characteristic
Quantity 50 g
UN Number UN1325
Sensitivity Air and moisture sensitive
Merck Index 14,4037
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 186.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ferrocene
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Ferrocene
Palabra de advertencia
  • Danger
Clasificación
  • Flammable solid Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'