Learn More
Ferroin Solution, Fisher Chemical™ Product Code.: 12920424
Ferroin Solution, Fisher Chemical™
Product Code. 12920424
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 12920424
Brand: Fisher Chemical RP 084 100
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1075967 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog NumberRP 084 100
Product Identifier
  • Ferroin Solution. European Pharmacopoeia Ref: 1038100 (RP 084)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : 1,10-phenanthrolinehydrochloride monohydrate, ferrous sulfate heptahydrate
Product Content Correction
Product Title
Ferroin Solution, Fisher Chemical™ >