missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate, 7-Hydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™
85.09 € - 421.00 €
Chemical Identifiers
CAS | 7782-63-0 |
---|---|
Molecular Formula | FeH14O11S |
Molecular Weight (g/mol) | 278.01 |
MDL Number | MFCD00149719 |
InChI Key | SURQXAFEQWPFPV-UHFFFAOYSA-L |
Synonym | iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate |
PubChem CID | 62662 |
ChEBI | CHEBI:75836 |
IUPAC Name | iron(2+);sulfate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15587954
|
Avantor J.T.Baker
2074-01 |
500 g | Wide Mouth Amber Glass Bottle | N/A | |||||
15597954
|
Avantor J.T.Baker
2074-05 |
2.5 kg | Wide Mouth Amber Glass Bottle |
421.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7782-63-0 | |
278.01 | |
SURQXAFEQWPFPV-UHFFFAOYSA-L | |
62662 | |
iron(2+);sulfate;heptahydrate |
FeH14O11S | |
MFCD00149719 | |
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
CHEBI:75836 | |
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
Specifications
7782-63-0 | |
FeH14O11S | |
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
278.01 | |
CHEBI:75836 | |
FCC/USP | |
Ferrous Sulfate, 7-Hydrate |
Crystals | |
MFCD00149719 | |
SURQXAFEQWPFPV-UHFFFAOYSA-L | |
iron(2+);sulfate;heptahydrate | |
62662 | |
278.02 | |
1L = 1.90kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title