missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Ferrules for Agilent™ Instruments
Ensure reliable connections for Agilent™ GC instruments with these Thermo Scientific™ Ferrules for Agilent Instruments.
Brand: Thermo Scientific™ 290GA139
43.33 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Use Thermo Scientific™ Ferrules for Agilent™ Instruments to ensure optimal performance of your Agilent GC instrument. They are available in two different materials of various dimension to accommodate a range of instruments and applications.
- Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
- Ferrules for different types of injectors and detectors
Specifications
Ferrule | |
Agilent Injectors and Non-MS Detectors | |
Pack of 10 |
0.1 to 0.32 mm | |
100% Graphite |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction