missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ferrules for Agilent™ Instruments

Ensure reliable connections for Agilent™ GC instruments with these Thermo Scientific™ Ferrules for Agilent Instruments.

Brand:  Thermo Scientific™ 290VA192

57.62 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10774827

  • 64.16 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ Ferrules for Agilent™ Instruments to ensure optimal performance of your Agilent GC instrument. They are available in two different materials of various dimension to accommodate a range of instruments and applications.

  • Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
  • Ferrules for different types of injectors and detectors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ferrule
0.32 mm
Agilent Injectors and Non-MS Detectors
15% Graphite, 85% Vespel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers