Learn More
Gibco™ Fetal Bovine Serum, charcoal stripped, USDA-approved regions
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.
91.50 € - 730.00 €
Specifications
Age | Fetal |
---|---|
Country of Origin | United States or Mexico |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Serum Treatment | Charcoal-stripped |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
15634559
|
Gibco™
A3382101 |
50 mL | One Shot |
91.50 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10706143
|
Gibco™
12676029 |
500 mL | Bottle |
730.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Serum Treatment: Stripped with charcoal-dextran
- Recommended Storage: -5°C to -20°C
Order Info
Shipping Condition: Dry Ice
Specifications
Fetal | |
Liquid | |
Charcoal-stripped |
United States or Mexico | |
Fetal Bovine Serum | |
Dry Ice |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.