missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, dialyzed, US origin
GREENER_CHOICE

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

111.00 € - 882.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin United States
Form Liquid
Product Type Fetal Bovine Serum
View More Specs

Products 2
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
15605639
GREENER_CHOICE
missing translation for 'viewDocuments'
Gibco™
A3382001
50 mL One Shot
111.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
11520646
GREENER_CHOICE
missing translation for 'viewDocuments'
Gibco™
26400044
500 mL Bottle
882.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Fetal Bovine Serum
Frozen
Fetal
Liquid
Dialyzed
Videos
SDS
Documents

Documents

Certificates

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.