missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, exosome-depleted
GREENER_CHOICE

Gibco™ Exosome-Depleted FBS streamLines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.

194.00 € - 1532.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin United States
Form Liquid
Product Type Fetal Bovine Serum
View More Specs

Products 2
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
15624559
GREENER_CHOICE
View Documents
Gibco™
A2720803
50 mL One Shot
194.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
15434573
GREENER_CHOICE
View Documents
Gibco™
A2720801
500 mL Bottle
1532.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco™ Exosome-Depleted FBS streamLines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.

  • Lowest exosome background
  • Enables consistent results: each lot tested for cell culture performance
  • No preparation needed: ultracentrifugation is not required

Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, for optimal assay results and increased experimental throughput.

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Fetal Bovine Serum
Sterile
Fetal
Liquid
Exosome-depleted
Frozen
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Fetal Bovine Serum, exosome-depleted

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.