missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions
GREENER_CHOICE

Gibco™ fetal bovine sera is specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.

112.00 € - 904.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Species Cattle/Bovine
Country of Origin Countries meeting USDA importation requirements
Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17887038
View Documents
Gibco™
12662002
50 mL
112.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
11538986
GREENER_CHOICE
View Documents
Gibco™
12662011
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11548986
View Documents
Gibco™
12662029
500 mL
904.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Origin: Countries meeting USDA importation requirements
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Storage: Store at -5°C to -20°C.

Specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Sterile
≤25 mg/dL
Frozen
Fetal
Countries meeting USDA importation requirements
Bottle
Standard (Sterile-filtered)
<50 EU/mL
MS Cell-Qualified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.