Learn More
Gibco™ Fetal Bovine Serum, qualified, United States
Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
73.75 € - 698.00 €
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
15575309
|
Gibco™
A3160501 |
50 mL | One Shot |
73.75 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15585309
|
Gibco™
A3160502 |
10 x 50 mL | One Shot | N/A | |||||
11570636
|
Gibco™
26140087 |
100 mL | Bottle | N/A | |||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
Gibco Sera: Trusted Quality for Consistent Performance
Gibco Sera Category: Value Plus (Performance)
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: United States
Gibco Serum Delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
Fetal Bovine Serum | |
Sterile | |
≤10 EU/mL | |
Qualified |
Fetal | |
Liquid | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
≤25 mg/dL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.