Learn More
Gibco™ Fetal Bovine Serum, qualified, USDA-approved regions
Important Update: We've recently introduced Value Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified USDA origin and Brazil (South American) origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ A3160601
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Replacing with Fetal Bovine Serum, value
- Sera application: for cell culture with robust cell lines, good value for basic research
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Origin: countries meeting USDA importation requirements
Gibco serum delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Fetal | |
Mexico | |
One Shot | |
50 mL | |
Sterile | |
Fewer resources used and less waste | |
One Shot™ |
Fetal Bovine Serum, qualified, One Shot™ format, USDA-approved regions | |
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
Qualified | |
Frozen |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.