missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fetal Bovine Serum, Tet system approved, US origin
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems.
114.00 € - 902.00 €
Specifications
Classification | Premium |
---|---|
Content And Storage | Store at <-10°C. |
Age | Fetal |
Cell Type | Use for a broad range of cell types, especially sensitive cell lines |
Species | Cattle/Bovine |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
16846796
|
Gibco™
A4736101 |
50 mL | One Shot |
114.00 €
50mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16856796
|
Gibco™
A4736201 |
500 mL | Bottle |
902.00 €
500mL |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems. Cell-based assays that detect the presence of tetracycline and tetracycline analogs are used to select FBS that delivers the highest range of induction.
- Designed to ensure the absence of detectable tetracycline and its derivatives that may interfere with accurate regulation of gene expression
- Functional testing provides confidence that unintended tetracyline gene expression will not result
- Sera Category: Specialty
Specifications
Premium | |
Fetal | |
Cattle/Bovine | |
Tet-System Approved | |
Sterile | |
Meets USP/EP guidelines with up to 91 harmonized quality specification tests | |
Sustainable packaging | |
Premium |
Store at <-10°C. | |
Use for a broad range of cell types, especially sensitive cell lines | |
United States | |
Fetal Bovine Serum | |
Triple-filtered, 0.1 μm | |
<5 EU/mL | |
<15 mg/dL | |
Frozen/Dry Ice |