missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, ultra-low IgG, US origin, One Shot™ format
GREENER_CHOICE

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

108.00 € - 884.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin United States
Form Liquid
Product Type Fetal Bovine Serum
View More Specs

Products 2
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
15695629
GREENER_CHOICE
View Documents
Gibco™
A3381901
50 mL One Shot
108.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
11550526
GREENER_CHOICE
View Documents
Gibco™
16250078
500 mL Bottle
884.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

IgG levels are less than 5 μg/mL, and the BVD titer is low or not detectable.

This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Fetal Bovine Serum
Fetal
Liquid
Frozen
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.