missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Value (formerly USDA-approved in North America or qualified, Brazil in other regions)

Gibco fetal bovine sera offers an excellent value for basic cell culture, specialty research, and specific assays. It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements.

49.08 € - 461.00 €

Specifications

Content And Storage Store at <-10°C.
Age Fetal
Species Cattle/Bovine
Country of Origin Refer to CoA for specific origin
Product Type Fetal Bovine Serum
View More Specs

Products 3
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
17553595
View Documents
Gibco™
A5209401
50 mL One Shot
49.08 €
50mL
Please to purchase this item. Need a web account? Register with us today!
17563595
View Documents
Gibco™
A5209402
10 x 50 mL One Shot
461.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
17479633
View Documents
Gibco™
A5256701
500 mL Bottle
389.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gibco Serum Delivers

  • ISO 13485-certified, processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in hood, reduces risk of contamination, and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

All products may not be available in all regions due to importation regulations

US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that help you meet your research needs and budget requirements.

  • Sera category: Value
  • Sera application: Cell culture with robust cell lines, good value for basic research
  • Testing: Up to 30 quality tests
  • Endotoxin level: ≤20 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: Refer to CoA for specific origin
Specifications

Specifications

Store at <-10°C.
Cattle/Bovine
Fetal Bovine Serum
Triple-filtered, 0.1 μm
≤20 EU/mL
≤25 mg/dL
Dry Ice
Fetal
Refer to CoA for specific origin
Sterile
Up to 30 quality tests including 9CFR virus testing* endotoxin, performance (*Please note: If manufactured in the UK, Full 9CFR except for Rabies and BlueTongue Virus which is tested via EMA)
Less waste
Value
USDA Safety Testing Performed for Mexican and Australian origins
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4 Years

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.