missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin-A Mouse anti-Human, Clone: 1F6B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66094-1-IG-150UL
This item is not returnable.
View return policy
Description
Human special AT-rich sequence-binding protein-2 (SATB2) is a nuclear matrix/scaffold-associated region DNA-binding protein. Like its homolog SATB1, SATB2 selectively binds double-stranded, special AT-rich DNA sequences, but is expressed primarily in a subset of postmitotic, differentiating neurons in the neocortex. Mice deficient in SATB exhibit craniofacial abnormalities and defects in osteoblast differentiation and function. SATB2 also interacts with and enhances the activity of Runx2 and ATF4, two transcription factors that regulate osteoblast differentiation, indicating that SATB2 acts as a molecular node in a transcriptional network regulating skeletal development and osteoblast differentiation. Recent experiments have shown that SATB2 interacts with histone deacetylase 1 and metastasis-associated protein 2, two proteins that are involved in chromatin remodeling, suggesting that SATB2 may also be involved in mediating epigenetic influences during cortical development.Specifications
Fetuin-A | |
Monoclonal | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02765 | |
AHSG | |
Fetuin-A Fusion Protein Ag10073 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
1F6B9 | |
Unconjugated | |
AHSG | |
A2HS, AHS, AHSG, alpha 2 HS glycoprotein, Alpha 2 Z globulin, Ba alpha 2 glycoprotein, FETUA, Fetuin A, HSGA | |
Mouse | |
Protein A | |
RUO | |
197 | |
-20°C | |
Liquid |