missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEV Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | FEV |
---|---|
Concentration | 0.45 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883595
|
Proteintech
25058-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873595
|
Proteintech
25058-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations.Specifications
FEV | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8QZW2, Q99581 | |
260298, 54738 | |
FEV Fusion Protein Ag20647 | |
Primary | |
-20°C | |
FEV |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FEV, FEV (ETS oncogene family), Fifth Ewing variant protein, HSRNAFEV, PC12 ETS factor 1, PET 1, PET1, Protein FEV | |
FEV | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |