missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FFAR2 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19952-1-AP-150UL
This item is not returnable.
View return policy
Description
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.Specifications
FFAR2 | |
Polyclonal | |
Unconjugated | |
FFAR2 | |
FFA2R, FFAR2, free fatty acid receptor 2, G protein coupled receptor 43, GPR43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100126285, 2867 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15552 | |
FFAR2 | |
Peptide | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG |