missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13784-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
FGF12 | |
Polyclonal | |
Unconjugated | |
FGF12 | |
FGF 12, FGF12, FGF12B, FHF 1, FHF1, fibroblast growth factor 12, Myocyte activating factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14167, 2257 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61328, P61329 | |
FGF12 | |
FGF12 Fusion Protein Ag4797 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction