missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIA Aromatics Standard ASTM D 1319 (Nominal value: 15.18% vol), Paragon Scientific™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- Certified Reference Material for Fluorescent Indicator Adsorption
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified via Round Robin
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
Specifications
| UN Number | UN 1223 |
| Quantity | 250 mL |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Kerosine sweetened
Nominal values quoted. Certified Values will vary from batch to batch.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction