missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fiacitabine, MedChemExpress
MedChemExpress Fiacitabine(NSC 382097; FIAC; FOAC) is a selective inhibitior of DNA replication of herpes simplex virus(HSV) with IC50 values of 2.5 nM and 12.6 nM for HSV1 and HSV2, respectively.
557.55 € - 796.50 €
Specifications
Chemical Name or Material | Fiacitabine |
---|---|
CAS | 69123-90-6 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C9H11FIN3O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16460558
|
Medchem Express
HY-50735 |
5 mg |
557.55 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16438098
|
Medchem Express
HY-50735 |
10 mg |
796.50 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fiacitabine(NSC 382097Specifications
Fiacitabine | |
Off-White | |
C9H11FIN3O4 | |
DMSO : ≥ 37 mg/mL (99.70 mM) | |
371.1 | |
98.83% | |
Solid |
69123-90-6 | |
Research | |
NSC 382097 FIAC FOAC | |
O=C(N=C1N)N([C@@H]([C@H]([C@@H]2O)F)O[C@@H]2CO)C=C1I | |
371.1 | |
Research | |
COVID-19-anti-virus |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only