missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Flexible Light Guide
Fiber Optic Flexible Light Guide in a black Megolon jacketing.
108.00 € - 194.00 €
Product Code | Brand | Type | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Length (Metric) | Price | Quantity & Availability | |||||
11706728
|
SCHOTT AG Lighting and Imaging™
121101 |
Single Fiber Optic Light Guide | 600 mm |
108.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
15753032
|
SCHOTT AG Lighting and Imaging™
171101 |
Single Fiber Optic Light Guide | 1000 mm |
132.00 €
1 unit |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
15763032
|
SCHOTT AG Lighting and Imaging™
171202 |
Double Fiber Optic Lightguide | 1000 mm |
194.00 €
1 unit |
In Stock
Log in to see stock. ![]() |
|||||
Description
SCHOTT Flexible Light Guides are highly bendable. In contrast to goosenecks, they allow a higher flexibility and need to be held.
- Light guide with eco-friendly Puravis fiber optics.
- Available in different lengths.
- Available with one or two branches.
- High white light transmission thanks to the use of SCHOTT PURAVIS™ fibers.
- High flexibility for different lighting, as well as remote illumination.
Specifications
Stereo Microscopy Illumination |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCHOTT AG Lighting and Imaging™ Fiber Optic Flexible Light Guide