Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Combi Light Guide
Fiber optic flex and stay double light guide in a black lacquered semi-rigid gooseneck tubing with an additional 800 mm fiber optic flexible light guide in a black metal-PVC jacketing.
Brand:  SCHOTT AG Lighting and Imaging™ 170402
Product Code. 15743032
  • 489.00 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Light guide with eco-friendly Puravis fiber optics.
  • Precise positioning offers complete control of light placement.
  • The combination of a double gooseneck light guide 4.5 mm dia. x 500 mm and a flexible light guide 4.5 mm dia. x 800 mm allows it to move further from the lightsource.
  • Active Fiber optic bundle: 2 x 4.5 mm for Gooseneck Outer Jacketing, 11 mm for gooseneck, and 14 mm for flexible.
  • Can be stationary fixed at the microscope post with the adapter 158 402.
  • To use with Cold light source KL1500HAL, KL1600LED or KL2500LED.
Specifications
Specifications
Fiber Optic Lightguide
Stereo Microscopy Illumination
2 x 500 mm for gooseneck and 800 mm for flexible
11 mm (Gooseneck), 14 mm (Flexible)
Cold Light Source KL1600LED, KL2500LED or KL1500HAL
Eco-friendly Puravis Glass Optical Fibers