Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand:  Thermo Scientific™ 5960
 View more versions of this product
Product Code. 15810530
  • 32.86 € / Pack of 12
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 Pack
64 (8 x 8 Grid)