Learn More
Brady™ Fiberglass-Reinforced Nylon Miniature Locking System for Circuit Breakers
A safe and effective method for locking out European Circuit Breakers.
Brand:  Brady™ 090848
 View more versions of this product
Product Code. 15603517
  • 129.00 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Specifications
Miniature Locking System for Circuit Breakers
Red/Black
Miniature Circuit Breaker Lockouts - Pin-In Standard
13.72 x 24.13 x 41.7 mm
6 Pk.
ELEC. CIRCUIT BREAKER L/O EURO PIS PK6
Fiberglass-reinforced Nylon
24.13 mm
Pin In
24.13 mm