Learn More
Thermo Scientific™ Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
GREENER_CHOICE
Separate up to fourteen 50 mL conical tubes at one time in a chemically-resistant and lighter option than aluminum with fixed angle rotors. They also offer safe alternatives in a repairable rotor with 15-year warranty to save on replacement costs.
Brand:  Thermo Scientific™ 75003661
4435.01 EUR valid until 2024-12-31
Use promo code "21988" to get your promotional price.
 View more versions of this product
Product Code. 10450655
  • 5487.30 € / Each
Explore more special offers
Description
Description
Separations of up to fourteen 50mL conical tubes at one time in a chemically-resistant and lighter option than aluminum. Thermo Scientific Fiberlite Rotors also offer safe alternatives in a repairable rotor with 15 year warranty to save on replacement costs.
Features:
Fiberlite Rotors:
  • Reduces run times with higher G-forces achieved with our lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design for structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Ordering Information:
Tube adapters are sold separately. For O-ring refresher kit, order 021-149-027 or contact your sales representative for more information.
Compatible with:
Thermo Scientific™ Multifuge™ X3/X3R/X3F/X3FR, Sorvall™ Legend™ XT/XF/XTR Centrifuge Series.
Recommended for:
Pelleting prokaryotic and eukaryotic cells from cell cultures.
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fiberlite F13-14 x 50cy Fixed Angle Rotor for Sorvall Legend X1/XT/XF
14 x 50 mL
9,250 rpm (10,000 rpm with 230V model)
1808
30 mm
34°
Energy efficient, less waste
Fixed Angle Rotor
14,636 x g (17,105 x g with 230 V)
8.3 cm
121 mm
Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Multifuge X3/X3F, Multifuge X1 and SL 40F Centrifuges
Yes