Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Brand:  Thermo Scientific™ 00456
 View more versions of this product
Product Code. 12796947
  • 587.93 € / Each
Explore more special offers
Description
Description
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:
  • Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7mL Round-bottom Tube