Learn More
Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor
GREENER_CHOICE
Separate samples safely at maximal g-force using the fixed angle rotor with bioseal technology.
Brand:  Thermo Scientific™ 46923
 View more versions of this product
Product Code. 12141680
  • 10954.50 € / Each
Explore more special offers
Description
Description
Safely separate samples at maximal g-force using the Thermo Scientific™ Fiberlite™ F21-8 x 50y Fixed-Angle Rotor with Bioseal technology.
Features:
Fiberlite Rotors:
  • Reduce run times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Compatible with:
Thermo Scientific Sorvall™ RC-6 Plus Series, Thermo Scientific™ Jouan KR22i centrifuge, Beckman™ J2 Series, Thermo Scientific™ Nalgene™ bottles and tubes
Recommended for:
Subcellular fractions, bacteria pelleting, lysate clarification, cell membranes, sucrose gradient isolation of mitochondria, endoplasmic reticulum and gradient separation of cells with Percoll, Ficoll, Optiprep or Nycodenz solutions
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fiberlite F21-8 x 50y Fixed-Angle Rotor
Fixed Angle Rotor
52,400 x g
106 mm
669
29 mm
Thermo Scientific™ Sorvall™ RC 6 Plus Centrifuge
16,299 x g
Energy efficient, less waste
Carbon Fiber Composite
8 x 50 mL
21,000 rpm
33 mm
115 mm
9.9 lb. (4.49 kg)
34°
Yes