missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 15 mL Conical Tube Adapters for Fiberlite F13-14 x 50cy Rotor and F15-8 x 50cy Rotor

Adapter for centrifuge rotor

Brand:  Thermo Scientific™ 75100378

217.41 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


Product Code. 10556434

  • 257.28 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Incorporate Thermo Scientific™ Fiberlite™ Rotor Adapters with Fiberlite F13-14x50c and F15-8x50c rotors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rotor Adapter
15 mL
15 mL
Conical
Adapter for 15mL conical tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.