Learn More
Thermo Scientific Chemicals Fibrinogen Related Peptide
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J66069.MA
558.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
HPLC | |
Solid | |
Fibrinogen Related Peptide | |
RUO - research use only |
-30°C to -10°C | |
1502.59 | |
10 mg | |
Gly-Gln-Gln-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only