missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fibronectin-Binding Protein
For research use
161.00 € - 259.00 €
Specifications
Formulation | Lyophilized powder |
---|---|
Storage Requirements | -30° to -10°C |
Regulatory Status | RUO |
Product Type | Fibronectin-Binding Protein |
Recombinant | Native |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498449
|
Thermo Scientific Chemicals
J66903.!D |
0.5mg |
161.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15408459
|
Thermo Scientific Chemicals
J66903.M^ |
1mg |
259.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lyophilized powder | |
RUO | |
Native |
-30° to -10°C | |
Fibronectin-Binding Protein | |
Solid |
RUO – Research Use Only