Learn More
Tocris Bioscience™ Filipin III
Polyene antibiotic; inhibits formation of pathological isoform of prion protein
Brand:  Tocris Bioscience™ 6250/1
Product Code. 18794858
  • 272.00 € / 1mg
Explore more special offers
Description
Description
Filipin III is a polyene antibiotic. Binds sterols in fungal membranes. Also inhibits formation of the pathological form of prion protein (PrP-res) from the normal membrane-bound (PrP-sen) form. Also fluorescent cholesterol stain.
Specifications
Specifications
Inhibitor
C35H58O11
Store at -20°C
1 mg
480-49-9
654.83g/mol
Antibiotics