Learn More
Buerkle™ Filling Support for Compact Jerrycan
Shop All Bürkle Products
Click to view available options
Non-sterile
Description
- Stainless steel AISI 304 (1.4301)
- For Bürkle compact jerrycans 10 litres (PP, transparent or HDPE, black, antistatic) )
- Scope of delivery: Filling support (consisting of holder and dispensing stand), mountable plastic feet
Specifications
Specifications
Autoclavable | Autoclavable |
For Use With (Equipment) | BüRkle Compact Jerry Can 10 L |
Material | Stainless Steel |
Sterility | Non-sterile |
Product Content Correction
Product Title
Buerkle™ Filling Support for Compact Jerrycan >
Spot an opportunity for improvement?Share a Content Correction