missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Filling Tubes for BRAND Seripettor, Dispensette S, Dispensette III and Dispensette Organic Bottletop Dispensers

Filling tubes for all BRAND Seripettor and Dispensette models made after 1997

16.55 € - 34.05 €


Products 4
Product Code Brand Description For Use With (Equipment) Material Length (Metric) Price Quantity & Availability  
Product Code Brand Description For Use With (Equipment) Material Length (Metric) Price Quantity & Availability  
15498013
View Documents
BRAND™
708210
Telescoping Filling Tubes For Nominal Volume 0.5, 1, 2, 5, 10 mL Dispensette™ S (all models), III, Organic, TA and seripettor™ pro FEP 70 to 140 mm
16.55 €
Each
In Stock
to see stock.
Add to basket
 
15418023
View Documents
BRAND™
708214
Telescoping Filling Tubes For Nominal Volume 0.5, 1, 2, 5, 10 mL Dispensette™ S (all models), III, Organic, TA and seripettor™ pro FEP 195 to 350 mm
33.10 €
Each
In Stock
to see stock.
Add to basket
 
15428023
View Documents
BRAND™
708216
Telescoping Filling Tubes For Nominal Volume 0.5, 1, 2, 5, 10 mL Dispensette™ S (all models), III, Organic, TA and seripettor™ pro FEP 250 to 480 mm
25.65 €
Each
In Stock
to see stock.
Add to basket
 
15448023
View Documents
BRAND™
708220
Telescoping Filling Tubes For Nominal Volume 25, 50, 100 mL Dispensette™ S (all models), III, Organic, TA and seripettor™ pro FEP 250 to 480 mm
34.05 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • 704536 - autoclavable filling tube for seripettor, 250mm, each
  • 704538 - autoclavable filling tube for seripettor, 500mm, each
  • Telescoping filling tubes adjust to correct height without cutting.
  • Unique design allows aspiration of liquid from bottom of bottle while minimizing aspiration of precipitates.
  • Two different diameter tubes: One for 1mL, 2mL, 5mL and 10mL nominal capacity dispensers; and one for 25mL, 50mL and 100mL nominal capacity dispensers
  • Five different lengths: 70-140mm; 125-240mm; 170-330mm; 195-350mm; 250-480mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Filling Tubes for BRAND Seripettor, Dispensette S, Dispensette III and Dispensette Organic Bottletop Dispensers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.