Learn More
Macherey-Nagel™ Pleated Filter MN 615 1/4
Use for chemical analysis in waste water treatment. Pleated Filter MN 615 1/4 is perfect for determination of dry matter.
Brand:  Macherey-Nagel™ 531032
 View more versions of this product
Product Code. 10423321
  • 69.40 € / Pack of 100
Explore more special offers
Description
Description
  • Chemical analysis in a waste water treatment plant
  • Ideally suited to be used for analytical procedures 
  • Reproducible results
  • Norm conforming filtration
  • Quality “Made in Germany”
  • Determination of dry matter - filtration for all N-parameters
Specifications
Specifications
Folded
Pleated Filter
320 mm
MN 615 1/4
320 mm