Learn More
Kartell™ Filtration and Vaccum Dessiccators
Can be evacuated to 1bar, with loss not exceeding 0.03mbar in 24 hours. Kartell™ Filtration and Vacuum Dessicators is implosion proof.
97.05 € - 151.00 €
Product Code | Brand | Height (Metric) Interior | Height (Metric) | Volume (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) Interior | Height (Metric) | Volume (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | |||
10365821
![]() |
Kartell™
0055000 |
135 mm | 190 mm | 2.15 L | 150 mm |
97.05 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||
10683272
![]() |
Kartell™
0023000 |
175 mm | 230 mm | 4.35 L | 200 mm |
143.00 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
10335921
![]() |
Kartell™
0055400 |
225 mm | 300 mm | 9.2 L | 250 mm |
151.00 €
Each |
In Stock
Log in to see stock. ![]() |
|||
Description
- Transparent polycarbonate cover seals onto a neoprene O-ring set into the flange of the polypropylene base
- Initial vacuum established with a little grease spread on the O-ring
- Removable pan for holding a drying agent
- Includes vacuum release tap/non-return valve assembly
- Specially grooved stopper in assembly allows air, when required, to re-enter very slowly to avoid disturbing the contents
Specifications
Polypropylene, Polycarbonate | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Kartell™ Filtration and Vaccum Dessiccators